Assay Details
| Gene Symbol: crn-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381579.1 | NP_001367040.1 | 2 - 3 | 1643 | 69 | |
| NM_001383937.1 | 4 - 5 | 2962 | 69 | |||
| GenBank mRNA | AY303577.1 | - | 2 - 3 | 1661 | 69 | |
| Gene Symbol: algn-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063881.6 | NP_496282.2 | 7 - 8 | 3791 | 69 | |
More Information
Set Membership: |
|

crn-3
Genomic Map