Assay Details
| Gene Symbol: crn-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063883.4 | NP_496284.1 | 1 - 2 | 219 | 160 | |
| GenBank mRNA | AY303579.1 | - | 1 - 2 | 209 | 160 | |
More Information
Set Membership: |
|

crn-5
Genomic Map