Assay Details
| Gene Symbol: mpz-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026867.4 | NP_001022038.2 | 22 - 23 | 2921 | 68 | |
| NM_001083157.4 | NP_001076626.2 | 16 - 17 | 2314 | 68 | ||
| NM_001129127.4 | NP_001122599.1 | 15 - 16 | 2260 | 68 | ||
| NM_001129129.4 | NP_001122601.1 | 20 - 21 | 2743 | 68 | ||
| NM_001377853.2 | NP_001364655.1 | 23 - 24 | 2974 | 68 | ||
| NM_001377854.2 | NP_001364657.1 | 22 - 23 | 2887 | 68 | ||
More Information
Set Membership: |
|

mpz-1
Genomic Map