Assay Details
| Gene Symbol: mpz-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026867.4 | NP_001022038.2 | 25 - 26 | 3234 | 69 | |
| NM_001083157.4 | NP_001076626.2 | 19 - 20 | 2627 | 69 | ||
| NM_001129127.4 | NP_001122599.1 | 18 - 19 | 2573 | 69 | ||
| NM_001129129.4 | NP_001122601.1 | 23 - 24 | 3056 | 69 | ||
| NM_001267333.3 | NP_001254262.1 | 2 - 3 | 248 | 69 | ||
| NM_001377853.2 | NP_001364655.1 | 26 - 27 | 3287 | 69 | ||
| NM_001377854.2 | NP_001364657.1 | 25 - 26 | 3200 | 69 | ||
More Information
Set Membership: |
|

mpz-1
Genomic Map