Assay Details
| Gene Symbol: mpz-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026867.4 | NP_001022038.2 | 33 - 34 | 5482 | 81 | |
| NM_001083157.4 | NP_001076626.2 | 27 - 28 | 4875 | 81 | ||
| NM_001129127.4 | NP_001122599.1 | 26 - 27 | 4821 | 81 | ||
| NM_001129129.4 | NP_001122601.1 | 31 - 32 | 5304 | 81 | ||
| NM_001267333.3 | NP_001254262.1 | 10 - 11 | 2496 | 81 | ||
| NM_001267334.3 | NP_001254263.1 | 3 - 4 | 1209 | 81 | ||
| NM_001377853.2 | NP_001364655.1 | 34 - 35 | 5535 | 81 | ||
| NM_001377854.2 | NP_001364657.1 | 33 - 34 | 5448 | 81 | ||
More Information
Set Membership: |
|

mpz-1
Genomic Map