Assay Details
| Gene Symbol: mpz-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026867.4 | NP_001022038.2 | 36 - 37 | 5858 | 87 | |
| NM_001083157.4 | NP_001076626.2 | 30 - 31 | 5251 | 87 | ||
| NM_001129127.4 | NP_001122599.1 | 29 - 30 | 5197 | 87 | ||
| NM_001129129.4 | NP_001122601.1 | 34 - 35 | 5680 | 87 | ||
| NM_001267333.3 | NP_001254262.1 | 13 - 14 | 2872 | 87 | ||
| NM_001267334.3 | NP_001254263.1 | 6 - 7 | 1585 | 87 | ||
| NM_001267335.3 | NP_001254264.1 | 1 - 2 | 55 | 87 | ||
| NM_001377853.2 | NP_001364655.1 | 37 - 38 | 5911 | 87 | ||
| NM_001377854.2 | NP_001364657.1 | 36 - 37 | 5824 | 87 | ||
More Information
Set Membership: |
|

mpz-1
Genomic Map