Assay Details
| Gene Symbol: mpz-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026867.4 | NP_001022038.2 | 13 - 14 | 1370 | 75 | |
| NM_001026868.5 | NP_001022039.2 | 13 - 14 | 1351 | 75 | ||
| NM_001083157.4 | NP_001076626.2 | 7 - 8 | 763 | 75 | ||
| NM_001129127.4 | NP_001122599.1 | 6 - 7 | 709 | 75 | ||
| NM_001129129.4 | NP_001122601.1 | 11 - 12 | 1192 | 75 | ||
| NM_001377853.2 | NP_001364655.1 | 14 - 15 | 1423 | 75 | ||
| NM_001377854.2 | NP_001364657.1 | 13 - 14 | 1336 | 75 | ||
More Information
Set Membership: |
|

mpz-1
Genomic Map