Assay Details
| Gene Symbol: ect-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063917.6 | NP_496318.1 | 10 - 11 | 1892 | 86 | |
| NM_063918.3 | NP_496319.1 | 11 - 12 | 1917 | 86 | ||
More Information
Set Membership: |
|

ect-2
Genomic Map