Assay Details
| Gene Symbol: gmps-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027085.4 | NP_001022256.2 | 4 - 5 | 751 | 71 | |
| NM_001027086.5 | NP_001022257.2 | 4 - 5 | 763 | 71 | ||
| NM_001083162.5 | NP_001076631.1 | 1 - 2 | 526 | 71 | ||
More Information
Set Membership: |
|

gmps-1
Genomic Map