Assay Details
| Gene Symbol: cap-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063935.9 | NP_496336.1 | 3 - 4 | 279 | 64 | |
| GenBank mRNA | Z18854.1 | - | 123 | 64 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cap-2| Gene Symbol: cap-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063935.9 | NP_496336.1 | 3 - 4 | 279 | 64 | |
| GenBank mRNA | Z18854.1 | - | 123 | 64 | ||
Set Membership: |
|