Assay Details
| Gene Symbol: cyn-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063936.7 | NP_496337.1 | 1 - 2 | 87 | 71 | |
| GenBank mRNA | AF421146.1 | - | 1 - 2 | 85 | 71 | |
| U36187.1 | - | 1 - 2 | 82 | 71 | ||
More Information
Set Membership: |
|

cyn-4
Genomic Map