Assay Details
| Gene Symbol: cyn-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063936.7 | NP_496337.1 | 3 - 4 | 990 | 126 | |
| GenBank mRNA | AF421146.1 | - | 3 - 4 | 988 | 126 | |
| U36187.1 | - | 3 - 4 | 985 | 126 | ||
More Information
Set Membership: |
|

cyn-4
Genomic Map