Assay Details
| Gene Symbol: orc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063938.5 | NP_496339.1 | 4 - 5 | 1107 | 134 | |
| GenBank mRNA | U40270.1 | - | 4 - 5 | 1116 | 134 | |
More Information
Set Membership: |
|

orc-2
Genomic Map