Assay Details
| Gene Symbol: orc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063938.5 | NP_496339.1 | 5 - 6 | 1187 | 83 | |
| GenBank mRNA | U40270.1 | - | 5 - 6 | 1196 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

orc-2| Gene Symbol: orc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063938.5 | NP_496339.1 | 5 - 6 | 1187 | 83 | |
| GenBank mRNA | U40270.1 | - | 5 - 6 | 1196 | 83 | |
Set Membership: |
|