Assay Details
| Gene Symbol: gly-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063944.7 | NP_496345.1 | 7 - 8 | 1548 | 71 | |
| GenBank mRNA | AY037793.1 | - | 1 - 2 | 263 | 71 | |
More Information
Set Membership: |
|

gly-1
Genomic Map