Assay Details
| Gene Symbol: gly-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063944.7 | NP_496345.1 | 9 - 10 | 1930 | 96 | |
| GenBank mRNA | AY037793.1 | - | 3 - 4 | 645 | 96 | |
More Information
Set Membership: |
|

gly-1
Genomic Map