Assay Details
| Gene Symbol: gly-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063944.7 | NP_496345.1 | 13 - 14 | 2611 | 65 | |
| GenBank mRNA | AY037793.1 | - | 7 - 8 | 1326 | 65 | |
More Information
Set Membership: |
|

gly-1
Genomic Map