Assay Details
| Gene Symbol: sra-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063947.2 | NP_496348.2 | 3 - 4 | 656 | 160 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sra-11| Gene Symbol: sra-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063947.2 | NP_496348.2 | 3 - 4 | 656 | 160 | |
Set Membership: |
|