Assay Details
| Gene Symbol: tceb-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063954.7 | NP_496355.1 | 4 - 5 | 1168 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tceb-3| Gene Symbol: tceb-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063954.7 | NP_496355.1 | 4 - 5 | 1168 | 75 | |
Set Membership: |
|