Assay Details
| Gene Symbol: mat-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063982.4 | NP_496383.2 | 6 - 7 | 3458 | 133 | |
| Gene Symbol: apc-17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063980.3 | NP_496381.1 | 9 - 10 | 4459 | 133 | |
More Information
Set Membership: |
|

mat-2
Genomic Map