Assay Details
| Gene Symbol: mat-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063982.4 | NP_496383.2 | 10 - 11 | 4443 | 93 | |
| Gene Symbol: apc-17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063980.3 | NP_496381.1 | 13 - 14 | 5444 | 93 | |
More Information
Set Membership: |
|

mat-2
Genomic Map