Assay Details
| Gene Symbol: ptr-13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063991.3 | NP_496392.2 | 8 - 9 | 2280 | 118 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ptr-13| Gene Symbol: ptr-13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063991.3 | NP_496392.2 | 8 - 9 | 2280 | 118 | |
Set Membership: |
|