Assay Details
| Gene Symbol: fndc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064003.7 | NP_496404.1 | 1 - 2 | 179 | 62 | |
| Gene Symbol: cchl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064002.7 | NP_496403.1 | 1 - 2 | 181 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_064002.7 transcripts.
Set Membership: |
|

fndc-1
Genomic Map