Assay Details
| Gene Symbol: rsf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027190.5 | NP_001022361.1 | 4 - 5 | 381 | 72 | |
| NM_001383950.1 | 4 - 5 | 391 | 72 | |||
| Gene Symbol: samp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064015.6 | NP_496416.1 | 10 - 11 | 3175 | 72 | |
More Information
Set Membership: |
|

rsf-1
Genomic Map