Assay Details
| Gene Symbol: shn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026835.6 | NP_001022006.1 | 10 - 11 | 3177 | 70 | |
| NM_001383954.1 | 11 - 12 | 3269 | 70 | |||
More Information
Set Membership: |
|

shn-1
Genomic Map