Assay Details
| Gene Symbol: shn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026835.6 | NP_001022006.1 | 2 - 3 | 409 | 95 | |
| NM_001026836.6 | NP_001022007.1 | 2 - 3 | 512 | 95 | ||
| NM_001383954.1 | 2 - 3 | 411 | 95 | |||
More Information
Set Membership: |
|

shn-1
Genomic Map