Assay Details
| Gene Symbol: shn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026835.6 | NP_001022006.1 | 3 - 4 | 513 | 63 | |
| NM_001026836.6 | NP_001022007.1 | 3 - 4 | 616 | 63 | ||
| NM_001383954.1 | 3 - 4 | 515 | 63 | |||
More Information
Set Membership: |
|

shn-1
Genomic Map