Assay Details
| Gene Symbol: shn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026835.6 | NP_001022006.1 | 8 - 9 | 2885 | 100 | |
| NM_001026836.6 | NP_001022007.1 | 8 - 9 | 2988 | 100 | ||
| NM_001383954.1 | 9 - 10 | 2977 | 100 | |||
More Information
Set Membership: |
|

shn-1
Genomic Map