Assay Details
| Gene Symbol: shn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026835.6 | NP_001022006.1 | 9 - 10 | 3034 | 85 | |
| NM_001383954.1 | 10 - 11 | 3126 | 85 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

shn-1| Gene Symbol: shn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026835.6 | NP_001022006.1 | 9 - 10 | 3034 | 85 | |
| NM_001383954.1 | 10 - 11 | 3126 | 85 | |||
Set Membership: |
|