Assay Details
| Gene Symbol: adbp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064038.8 | NP_496439.1 | 2 - 3 | 208 | 106 | |
| GenBank mRNA | AF303256.1 | - | 2 - 3 | 208 | 106 | |
More Information
Set Membership: |
|

adbp-1
Genomic Map