Assay Details
| Gene Symbol: ldh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001356894.2 | NP_001343759.1 | 3 - 4 | 885 | 78 | |
| NM_064102.6 | NP_496503.1 | 2 - 3 | 839 | 78 | ||
| GenBank mRNA | U15420.1 | - | 2 - 3 | 838 | 78 | |
More Information
Set Membership: |
|

ldh-1
Genomic Map