Assay Details
| Gene Symbol: alh-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026907.4 | NP_001022078.1 | 1 - 2 | 56 | 54 | |
| NM_001026908.6 | NP_001022079.1 | 1 - 2 | 62 | 54 | ||
More Information
Set Membership: |
|

alh-8
Genomic Map