Assay Details
| Gene Symbol: alh-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026907.4 | NP_001022078.1 | 4 - 5 | 792 | 138 | |
| NM_001026908.6 | NP_001022079.1 | 4 - 5 | 808 | 138 | ||
More Information
Set Membership: |
|

alh-8
Genomic Map