Assay Details
| Gene Symbol: ash-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373798.2 | NP_001360415.1 | 2 | 175 | 80 | |
| NM_001373799.3 | NP_001359515.1 | 2 - 3 | 214 | 80 | ||
| NR_138517.1 | 2 - 3 | 802 | 80 | |||
More Information
Set Membership: |
|

ash-2
Genomic Map