Assay Details
| Gene Symbol: ash-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373799.3 | NP_001359515.1 | 4 - 5 | 767 | 97 | |
| NM_064154.3 | NP_496555.1 | 4 - 5 | 724 | 97 | ||
| NR_138517.1 | 4 - 5 | 1355 | 97 | |||
| GenBank mRNA | EU178102.1 | - | 4 - 5 | 712 | 97 | |
More Information
Set Membership: |
|

ash-2
Genomic Map