Assay Details
| Gene Symbol: ash-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001330962.4 | NP_001317781.1 | 6 - 7 | 1109 | 155 | |
| NM_001373798.2 | NP_001360415.1 | 6 | 1114 | 155 | ||
| NM_001373799.3 | NP_001359515.1 | 6 - 7 | 1144 | 155 | ||
| NM_064154.3 | NP_496555.1 | 6 - 7 | 1101 | 155 | ||
| NR_138517.1 | 7 - 8 | 1784 | 155 | |||
| GenBank mRNA | EU178102.1 | - | 6 - 7 | 1089 | 155 | |
More Information
Set Membership: |
|

ash-2
Genomic Map