Assay Details
| Gene Symbol: ash-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001330962.4 | NP_001317781.1 | 8 - 9 | 1418 | 79 | |
| NM_001373798.2 | NP_001360415.1 | 8 | 1423 | 79 | ||
| NM_001373799.3 | NP_001359515.1 | 8 - 9 | 1453 | 79 | ||
| NM_064154.3 | NP_496555.1 | 8 - 9 | 1410 | 79 | ||
| NR_138517.1 | 9 - 10 | 2093 | 79 | |||
| GenBank mRNA | EU178102.1 | - | 8 - 9 | 1398 | 79 | |
More Information
Set Membership: |
|

ash-2
Genomic Map