Assay Details
| Gene Symbol: ash-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001330962.4 | NP_001317781.1 | 2 - 3 | 175 | 74 | |
| NM_064154.3 | NP_496555.1 | 2 - 3 | 176 | 74 | ||
| GenBank mRNA | EU178102.1 | - | 2 - 3 | 164 | 74 | |
More Information
Set Membership: |
|

ash-2
Genomic Map