Assay Details
| Gene Symbol: mcm-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027245.8 | NP_001022416.1 | 2 - 3 | 1049 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mcm-2| Gene Symbol: mcm-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027245.8 | NP_001022416.1 | 2 - 3 | 1049 | 69 | |
Set Membership: |
|