Assay Details
| Gene Symbol: ras-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064222.7 | NP_496623.1 | 3 - 4 | 400 | 126 | |
| NR_051787.1 | 3 - 4 | 307 | 126 | |||
| GenBank mRNA | U56930.1 | - | 3 - 4 | 375 | 126 | |
More Information
Set Membership: |
|

ras-1
Genomic Map