Assay Details
| Gene Symbol: ras-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064222.7 | NP_496623.1 | 5 - 6 | 619 | 115 | |
| GenBank mRNA | U56930.1 | - | 5 - 6 | 594 | 115 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ras-1| Gene Symbol: ras-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064222.7 | NP_496623.1 | 5 - 6 | 619 | 115 | |
| GenBank mRNA | U56930.1 | - | 5 - 6 | 594 | 115 | |
Set Membership: |
|