Assay Details
Gene Symbol: dct-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064240.2 | NP_496641.2 | 6 - 7 | 882 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: dct-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064240.2 | NP_496641.2 | 6 - 7 | 882 | 66 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |