Assay Details
| Gene Symbol: T21B4.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064265.3 | NP_496666.1 | 2 - 3 | 862 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

T21B4.3| Gene Symbol: T21B4.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064265.3 | NP_496666.1 | 2 - 3 | 862 | 92 | |
Set Membership: |
|