Assay Details
| Gene Symbol: rgs-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001377886.2 | NP_001364555.1 | 2 - 3 | 242 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rgs-4| Gene Symbol: rgs-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001377886.2 | NP_001364555.1 | 2 - 3 | 242 | 75 | |
Set Membership: |
|