Assay Details
| Gene Symbol: rgs-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267421.4 | NP_001254350.3 | 3 - 4 | 354 | 78 | |
| NM_001373807.2 | NP_001360477.1 | 3 - 4 | 389 | 78 | ||
| NM_001377886.2 | NP_001364555.1 | 5 - 6 | 563 | 78 | ||
| NM_001381631.1 | NP_001368098.1 | 2 - 3 | 162 | 78 | ||
More Information
Set Membership: |
|

rgs-4
Genomic Map