Assay Details
| Gene Symbol: snf-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064334.5 | NP_496735.1 | 5 - 6 | 1988 | 64 | |
| GenBank mRNA | AY825249.1 | - | 5 - 6 | 1878 | 64 | |
More Information
Set Membership: |
|

snf-5
Genomic Map