Assay Details
| Gene Symbol: ptr-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064360.5 | NP_496761.3 | 7 - 8 | 1075 | 128 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ptr-18| Gene Symbol: ptr-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064360.5 | NP_496761.3 | 7 - 8 | 1075 | 128 | |
Set Membership: |
|