Assay Details
| Gene Symbol: max-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083166.5 | NP_001076635.1 | 8 - 9 | 1501 | 87 | |
| NM_001083167.2 | NP_001076636.1 | 4 - 5 | 1021 | 87 | ||
| NM_001129183.4 | NP_001122655.1 | 9 - 10 | 1651 | 87 | ||
| NM_001267434.3 | NP_001254363.1 | 2 - 3 | 454 | 87 | ||
| NM_001313556.2 | NP_001300485.1 | 9 - 10 | 1654 | 87 | ||
| NM_001313557.3 | NP_001300486.1 | 8 | 1450 | 87 | ||
| GenBank mRNA | DQ523832.1 | - | 8 - 9 | 1501 | 87 | |
More Information
Set Membership: |
|

max-2
Genomic Map