Assay Details
| Gene Symbol: F18A11.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064370.5 | NP_496771.1 | 5 - 6 | 1109 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F18A11.2| Gene Symbol: F18A11.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064370.5 | NP_496771.1 | 5 - 6 | 1109 | 74 | |
Set Membership: |
|