Assay Details
| Gene Symbol: gcn-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381636.1 | NP_001367229.1 | 1 - 2 | 130 | 92 | |
| NM_001381637.1 | NP_001366669.1 | 2 - 3 | 228 | 92 | ||
More Information
Set Membership: |
|

gcn-2
Genomic Map